Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102175315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058173.1 | XP_017913662.1 | 3 - 4 | 407 | 104 | |
GenBank mRNA | JO428430.1 | - | 3 - 4 | 596 | 104 | |
JO428442.1 | - | 3 - 4 | 596 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |