Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690631.3 | XP_005690688.2 | 1 - 2 | 238 | 91 | |
XM_018060293.1 | XP_017915782.1 | 1 - 2 | 238 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |