Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRARG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693295.3 | XP_005693352.2 | 2 - 3 | 848 | 114 | |
GenBank mRNA | JT746839.1 | - | 1 - 2 | 156 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |