Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695072.3 | XP_005695129.1 | 1 - 2 | 112 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |