Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697521.3 | XP_005697578.3 | 3 - 4 | 502 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |