Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699206.3 | XP_005699263.2 | 3 - 4 | 2516 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |