Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682498.3 | XP_005682555.2 | 6 - 7 | 2521 | 87 | |
GenBank mRNA | JO625515.2 | - | 1 - 2 | 424 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |