Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692755.3 | XP_005692812.2 | 2 - 3 | 167 | 103 | |
XM_005692756.3 | XP_005692813.1 | 2 - 3 | 156 | 103 | ||
XM_005692757.3 | XP_005692814.1 | 2 - 3 | 249 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |