Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063065.1 | XP_017918554.1 | 2 - 3 | 262 | 77 | |
GenBank mRNA | JO377005.2 | - | 2 - 3 | 237 | 77 | |
KA311333.1 | - | 4 - 5 | 797 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |