Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067113.1 | XP_017922602.1 | 2 - 3 | 2034 | 68 | |
GenBank mRNA | JO645739.2 | - | 2 - 3 | 1383 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |