Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043622.1 | XP_017899111.1 | 1 - 2 | 155 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |