Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682367.3 | XP_005682424.1 | 3 - 4 | 563 | 63 | |
XM_005682368.2 | XP_005682425.1 | 3 - 4 | 592 | 63 | ||
GenBank mRNA | JO433735.2 | - | 1 - 2 | 1186 | 63 | |
JO645961.2 | - | 1 - 2 | 1496 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |