Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055808.1 | XP_017911297.1 | 3 - 4 | 566 | 100 | |
GenBank mRNA | JO438826.2 | - | 2 - 3 | 610 | 100 | |
JO592986.2 | - | 3 - 4 | 557 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |