Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687669.3 | XP_005687726.1 | 4 - 5 | 519 | 76 | |
GenBank mRNA | JO340496.2 | - | 1 - 2 | 1525 | 76 | |
JO340497.2 | - | 1 - 2 | 1525 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |