Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964668.2 | XP_013820122.1 | 9 - 10 | 1215 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |