Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066470.1 | XP_017921959.1 | 3 - 4 | 2073 | 86 | |
XM_018066471.1 | XP_017921960.1 | 2 - 3 | 712 | 86 | ||
GenBank mRNA | JO403334.2 | - | 2 - 3 | 86 | ||
JO444625.2 | - | 1 - 2 | 264 | 86 | ||
KA339666.1 | - | 2 - 3 | 665 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |