Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975370.2 | XP_013830824.2 | 2 - 3 | 247 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |