Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043250.1 | XP_017898739.1 | 4 - 5 | 1011 | 77 | |
XM_018043251.1 | XP_017898740.1 | 4 - 5 | 927 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |