Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044592.1 | XP_017900081.1 | 5 - 6 | 2160 | 62 | |
XM_018044593.1 | XP_017900082.1 | 6 - 7 | 2404 | 62 | ||
XM_018044594.1 | XP_017900083.1 | 5 - 6 | 2278 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |