Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679809.2 | XP_005679866.1 | 2 - 3 | 1176 | 76 | |
XM_005679810.3 | XP_005679867.1 | 3 - 4 | 1015 | 76 | ||
GenBank mRNA | JO365016.2 | - | 2 - 3 | 1083 | 76 | |
JT651908.1 | - | 2 - 3 | 196 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |