Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10H15orf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685467.3 | XP_005685524.1 | 5 - 6 | 553 | 71 | |
XM_013966954.2 | XP_013822408.1 | 5 - 6 | 553 | 71 | ||
XM_013966957.2 | XP_013822411.1 | 6 - 7 | 592 | 71 | ||
XM_018054491.1 | XP_017909980.1 | 5 - 6 | 553 | 71 | ||
GenBank mRNA | JT711445.1 | - | 5 - 6 | 435 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |