Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040643.1 | XP_017896132.1 | 10 - 11 | 1100 | 67 | |
GenBank mRNA | JO426112.1 | - | 9 - 10 | 878 | 67 | |
JO426157.1 | - | 9 - 10 | 878 | 67 | ||
JO637366.2 | - | 10 - 11 | 1060 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |