Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040643.1 | XP_017896132.1 | 12 - 13 | 1333 | 75 | |
GenBank mRNA | JO426112.1 | - | 11 - 12 | 1111 | 75 | |
JO426157.1 | - | 11 - 12 | 1111 | 75 | ||
JO637366.2 | - | 12 - 13 | 1293 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |