Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO426112.1 | - | 1 - 2 | 215 | 95 | |
JO426157.1 | - | 1 - 2 | 215 | 95 | ||
JO637366.2 | - | 2 - 3 | 397 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |