Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040643.1 | XP_017896132.1 | 8 - 9 | 972 | 97 | |
GenBank mRNA | JO426112.1 | - | 7 - 8 | 750 | 97 | |
JO426157.1 | - | 7 - 8 | 750 | 97 | ||
JO637366.2 | - | 8 - 9 | 932 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |