Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051147.1 | XP_017906636.1 | 4 - 5 | 546 | 99 | |
XM_018051148.1 | XP_017906637.1 | 4 - 5 | 1574 | 99 | ||
XM_018051149.1 | XP_017906638.1 | 4 - 5 | 553 | 99 | ||
GenBank mRNA | JO439178.2 | - | 4 - 5 | 518 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |