Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053894.1 | XP_017909383.1 | 5 - 6 | 929 | 68 | |
GenBank mRNA | JO576630.1 | - | 5 - 6 | 806 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |