Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053929.1 | XP_017909418.1 | 4 - 5 | 919 | 65 | |
GenBank mRNA | JT739813.1 | - | 5 - 6 | 691 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |