Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20H5orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065515.1 | XP_017921004.1 | 6 - 7 | 1712 | 128 | |
XM_018065516.1 | XP_017921005.1 | 6 - 7 | 1600 | 128 | ||
XM_018065517.1 | XP_017921006.1 | 6 - 7 | 1657 | 128 | ||
XM_018065518.1 | XP_017921007.1 | 6 - 7 | 1545 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |