Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041575.1 | XP_017897064.1 | 5 - 6 | 1180 | 102 | |
GenBank mRNA | JO425207.1 | - | 2 - 3 | 799 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |