Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041575.1 | XP_017897064.1 | 7 - 8 | 1421 | 68 | |
GenBank mRNA | JO425207.1 | - | 4 - 5 | 1040 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |