Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C26H10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975309.2 | XP_013830763.2 | 2 - 3 | 771 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |