Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044424.1 | XP_017899913.1 | 2 - 3 | 265 | 78 | |
XM_018044425.1 | XP_017899914.1 | 2 - 3 | 1447 | 78 | ||
GenBank mRNA | JO440402.2 | - | 2 - 3 | 180 | 78 | |
JO555784.2 | - | 2 - 3 | 206 | 78 | ||
JO641394.2 | - | 1 - 2 | 501 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |