Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674988.3 | XP_005675045.2 | 2 - 3 | 453 | 83 | |
GenBank mRNA | JO375330.2 | - | 2 - 3 | 386 | 83 | |
JO557335.2 | - | 420 | 83 | |||
JO557336.2 | - | 2 - 3 | 405 | 83 | ||
JO612247.2 | - | 2 - 3 | 460 | 83 | ||
JO644795.2 | - | 2 - 3 | 547 | 83 | ||
JT673041.1 | - | 2 - 3 | 296 | 83 | ||
JT674050.1 | - | 1 - 2 | 83 | 83 | ||
JT684042.1 | - | 1 - 2 | 88 | 83 | ||
JT695587.1 | - | 2 - 3 | 207 | 83 | ||
JT726725.1 | - | 3 - 4 | 260 | 83 | ||
JT727518.1 | - | 1 - 2 | 76 | 83 | ||
JT750471.1 | - | 1 - 2 | 87 | 83 | ||
KA337235.1 | - | 2 - 3 | 368 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |