Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686002.3 | XP_005686059.1 | 4 - 5 | 701 | 91 | |
XM_013967249.2 | XP_013822703.2 | 5 - 6 | 856 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |