Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689554.3 | XP_005689611.2 | 1 - 2 | 785 | 67 | |
GenBank mRNA | JO429725.2 | - | 1 - 2 | 578 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |