Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695724.3 | XP_005695781.2 | 2 - 3 | 906 | 84 | |
XM_018038393.1 | XP_017893882.1 | 2 - 3 | 503 | 84 | ||
XM_018038394.1 | XP_017893883.1 | 2 - 3 | 272 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |