Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039636.1 | XP_017895125.1 | 2 - 3 | 299 | 84 | |
GenBank mRNA | JO636740.2 | - | 2 - 3 | 161 | 84 | |
JT751608.1 | - | 1 - 2 | 73 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |