Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THRSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO421667.2 | - | 1 - 2 | 831 | 86 | |
KA327386.1 | - | 1 - 2 | 451 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |