Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050596.1 | XP_017906085.1 | 3 - 4 | 565 | 115 | |
XM_018050597.1 | XP_017906086.1 | 3 - 4 | 599 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |