Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049157.1 | XP_017904646.1 | 6 - 7 | 917 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |