Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969538.2 | XP_013824992.1 | 3 - 4 | 708 | 77 | |
XM_018059657.1 | XP_017915146.1 | 3 - 4 | 499 | 77 | ||
GenBank mRNA | KA335885.1 | - | 1 - 2 | 1650 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |