Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061771.1 | XP_017917260.1 | 2 - 3 | 229 | 71 | |
GenBank mRNA | JO376441.2 | - | 2 - 3 | 267 | 71 | |
JO559743.2 | - | 2 - 3 | 1661 | 71 | ||
JO580947.1 | - | 2 - 3 | 1705 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |