Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972587.2 | XP_013828041.1 | 10 - 11 | 2092 | 90 | |
XM_013972588.2 | XP_013828042.1 | 8 - 9 | 1431 | 90 | ||
XM_013972589.2 | XP_013828043.1 | 7 - 8 | 975 | 90 | ||
XM_018064928.1 | XP_017920417.1 | 9 - 10 | 1494 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |