Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041182.1 | XP_017896671.1 | 6 - 7 | 790 | 63 | |
XM_018041183.1 | XP_017896672.1 | 6 - 7 | 760 | 63 | ||
XM_018041184.1 | XP_017896673.1 | 5 - 6 | 700 | 63 | ||
XM_018041185.1 | XP_017896674.1 | 5 - 6 | 691 | 63 | ||
XM_018041186.1 | XP_017896675.1 | 3 - 4 | 441 | 63 | ||
XM_018041188.1 | XP_017896677.1 | 4 - 5 | 529 | 63 | ||
GenBank mRNA | JT667525.1 | - | 6 - 7 | 830 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |