Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042907.1 | XP_017898396.1 | 3 | 2692 | 68 | |
GenBank mRNA | KA339248.1 | - | 2 | 1859 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |