Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4H7orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047000.1 | XP_017902489.1 | 2 | 1265 | 97 | |
GenBank mRNA | JO647708.2 | - | 1 | 567 | 97 | |
JT688293.1 | - | 2 | 1171 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |