Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044973.1 | XP_017900462.1 | 8 - 9 | 1080 | 77 | |
GenBank mRNA | JO424441.1 | - | 8 - 9 | 1157 | 77 | |
KA330779.1 | - | 2 - 3 | 1122 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |