Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692232.3 | XP_005692289.1 | 2 - 3 | 288 | 95 | |
GenBank mRNA | JO610184.2 | - | 5 - 6 | 984 | 95 | |
JO629273.2 | - | 4 - 5 | 389 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |