Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042748.1 | XP_017898237.1 | 3 - 4 | 557 | 69 | |
GenBank mRNA | JT668557.1 | - | 1 - 2 | 41 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |